EMD Millipore Calbiochem™ Caspase-6 Inhibitor I

The Caspase-6 Inhibitor I controls the biological activity of Caspase-6. This small molecule/inhibitor is primarily used for Cancer applications.

Manufacturer: EMD Millipore 2187571MG

 View more versions of this product

Catalog No. 21-875-71MG

  Check Availability
Add to Cart

    Specifications

    Storage Requirements Protect from Moisture
    Target Proteins caspase-6
    Molecular Formula C31H45FN4O10
    Formula Weight 652.7g/mol
    Solubility DMSO
    Description 90% by TLC
    Format Lyophilized solid
    Quantity 1mg

    • Peptide Sequence: Z-Val-Glu(OMe)-Ile-Asp(OMe)-CH2F*
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit