Learn More
EMD Millipore Calbiochem™ Caspase Inhibitor III
The Caspase Inhibitor III, also referenced under CAS 634911-80-1, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.
Manufacturer:  EMD Millipore 2187451MG
 View more versions of this product
Catalog No. 21-874-51MG
  Check Availability
Add to cart
Specifications
Description
  • Peptide Sequence: Boc-Asp(OMe)-CH2F*
  • Cell Permeable
Description & Specifications
Specifications
single spot by TLC
Protect from Moisture
broad spectrum caspase
C11H18FNO5
DMSO
Off-white semi-solid
1mg
634911-80-1
263.3g/mol