Learn More
EMD Millipore Calbiochem™ Caspase Inhibitor, Negative Control Available on GSA/VA Contract for Federal Government customers only.
The Caspase Inhibitor, Negative Control, also referenced under CAS 105637-38-5, controls the biological activity of Caspase. This small molecule/inhibitor is primarily used for Cancer applications.
Manufacturer:  EMD Millipore 3420005MG
 View more versions of this product
Catalog No. 34-200-05MG
  Check Availability
Add to cart
Specifications
Description
  • Peptide Sequence: Z-Phe-Ala-FMK
  • Cell Permeable
Description & Specifications
Specifications
98% by HPLC
Protect from Moisture
Cathepsin-B
C21H23FN2O4
DMSO
Off-white solid
5mg
105637-38-5
386.4g/mol