Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cathepsin S Substrate, Fluorogenic
Click to view available options
Quantity:
1 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: Mca-Gly-Arg-Trp-Pro-Pro-Met-Gly∼Leu-Pro-Trp-Glu-Lys(Dnp)-D-Arg-NH2

Specifications
Specifications
Color | Yellow |
Formulation | Supplied as a trifluoroacetate salt. |
Description | 95% by HPLC |
Molecular Formula | C93H123N25O23S |
Formula Weight | 1991.2g/mol |
Solubility | DMSO |
Content And Storage | Hygroscopic, Protect from light |
Form | Solid |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction