EMD Millipore Calbiochem™ Cdk2 Inhibitor II Available on GSA/VA Contract for Federal Government customers only.

The Cdk2 Inhibitor II, also referenced under CAS 222035-13-4, controls the biological activity of Cdk2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Manufacturer: EMD Millipore 2194455MG

 View more versions of this product

Catalog No. 21-944-55MG

  Check Availability
Add to Cart

    Specifications

    Format Yellow solid
    Solubility DMSO
    Description 95% by HPLC (sum of two isomers)
    Storage Requirements Protect from Light
    Target Proteins CDK2
    Molecular Formula C14H11BrN4O3S
    Formula Weight 395.2g/mol
    Quantity 5mg
    CAS 222035-13-4

    • Packaged Under Inert Gas
    • Cell Permeable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit