Learn More
EMD Millipore™ Calbiochem™ Cell Culture-Tested Hygromycin B, Streptomyces sp.
For identification or selection of recombinant clones in variety of cell types
Manufacturer:  EMD Millipore™ 400050100MG
 View more versions of this product
Catalog No. 40-005-0100MG
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
50μg/mL to 1mg/mL for mammalian cell selection
Hygromycin B
2- 8°C
Light brown lyophilized solid
100mg