Learn More
MilliporeSigma™ Calbiochem™ Collagen, Type II, Bovine Joint Cartilage
Isolated from triple helical domain of guanidine-HCl-extracted and pepsin-digested bovine joint cartilage collagen Type II
Supplier:  MilliporeSigma™ 2341841MG
Catalog No. 23-418-41MG
Add to Cart
Description
Description
  • Highly purified preparation
  • Does not react with anti-collagen Type I and anti-collagen Type III by Western blotting
Specifications
Specifications
1 mg
-20°C
≥95% by SDS-PAGE
Collagen, Bovine Joint Cartilage Type II
Lyophilized
Product Suggestions
Product Suggestions