Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ CRAC Channel Inhibitor IV, EVP4593
Click to view available options
Quantity:
5 mg
Description
Packaged Under Inert Gas

Specifications
Specifications
Color | Yellow |
Description | 98% by HPLC |
Molecular Formula | C22H20N4O |
Formula Weight | 356.42g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction