Learn More
MilliporeSigma™ Calbiochem™ CRM1 Inhibitor IV, KPT-251
A cell-permeable, irreversible, triazole-containing heterotricyclic CRM1 inhibitor that effectively blocks CRM1-mediated nuclear export (4 h 1μM in U2OS cells) via covalent interaction with CRM1 NES- (nuclear export signal) binding groove cysteine (Cys528 in human)
Supplier: MilliporeSigma™ 5.00505.0001
Description
A cell-permeable, irreversible, triazole-containing heterotricyclic CRM1 inhibitor that effectively blocks CRM1-mediated nuclear export (4 h 1μM in U2OS cells) via covalent interaction with CRM1 NES- (nuclear export signal) binding groove cysteine (Cys528 in human). Exhibits antileukemic activity against primary human CD19+ CLL (IC50 <500nM) via apoptosis induction even in the presence of stroma cells or other known CLL survival stimuli, while exhibiting little cytotoxicity toward CD56+ NK cells from healthy donors (1μM; up to 72 h). Reported to prolong the survival of SCID mice (75mg/kg p.o.) engrafted with murine Eμ-TCL1 or human MV4-11 leukemia in vivo .Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
Specifications
Solid | |
5 mg | |
99% by HPLC | |
C14H7F6N5O | |
DMSO |
Protect from light | |
White | |
XPO1 | |
375.23g/mol |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.