Learn More
MilliporeSigma™ Calbiochem™ CXCR2 antagonist, Cpd 19
The CXCR2 antagonist, Cpd 19, also referenced under CAS 473728-67-5, controls the biological activity of CXCR2.
Manufacturer:  MilliporeSigma™ 2398192MG
Catalog No. 23-981-92MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
95% by HPLC
Protect from Light
473728-67-5
343.38g/mol
Off-white solid
2mg
C18H21N3O4
DMSO