Learn More
MilliporeSigma™ Calbiochem™ Cyclophilin A, Human, Recombinant, E. coli
Recombiant, human cyclophilin A expressed in E.
Manufacturer:  MilliporeSigma™ 239777100UG
Catalog No. 23-977-7100UG
Add to cart
Description
Description
Recombiant, human cyclophilin A expressed in E. coli . An ubiquitous enzyme that catalyzes the cis-trans- isomerization of X-Pro peptide bonds. Binds to proteins and acts as a chaperone in protein folding. Incorporates into HIV-1 virons and is believed to be essential for viral infectivity. Cyclophilin binds to the immunosuppressant Cyclosporin A (Cat. No. 239835). This complex then binds calcineurin (PP2B) and inhibits its phosphatase activity.
Specifications
Specifications
In 20 mM Tris-HCl, pH 7.8.
Liquid
Catalyzes the cis-trans-isomerization of X-Pro peptide bonds
18000g/mol
90% by SDS-PAGE
100μg
95076-93-0