Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Cyclosporine A, Tolypocladium inflatum

Induces apoptosis in rat thymocytes, murine B cell lymphoma cell line, WEH1-231.

Manufacturer:  MilliporeSigma™ 239835100MG

Catalog No. 23-983-5100MG


Add to cart

Description

Description

EMD Millipore Calbiochem™ Cyclosporine A, Tolypocladium inflatum is Cyclic oligopeptide with immunosuppressant properties.

  • Prevents anti-IgM, ionomycin-induced apoptosis in BLB cell lines
  • Complex of cyclosporin A with cyclophilin inhibits protein phosphatase 2B with nanomolar affinity
  • Inhibits nitric oxide synthesis induced by interleukin-1α, lipopolysaccharides, TNF-α
  • Induces cardiomyocytes from embryonic stem cells
  • Assay: ≥97% (dry basis)
  • CAS: 59865-13-3
  • Molecular Formula: C62H11N11O12
  • Color: White to Off-White
Specifications

Specifications

2°C to 8°C
100 mg
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit