Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Cyclothiazide
Click to view available options
Quantity:
10 mg
Description
Selectively blocks AMPA receptor desensitization and potentiates AMPA and kainate-induced increases in intracellular calcium and norepinephrine release.

Specifications
Specifications
Color | White |
Description | 98% by HPLC (sum of two diastereoisomers) |
Target Proteins | AMPA receptor |
Molecular Formula | C14H16ClN3O4S2 |
Solubility | DMSO or EtOH |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction