Learn More
MilliporeSigma™ Calbiochem™ Daunorubicin, Hydrochloride
Potent cell-permeable anticancer agent whose potential target site may be mitochondrial cytochrome c oxidase.
Manufacturer:  MilliporeSigma™ 2518005MG
Catalog No. 25-180-05MG
Add to cart
Description
Description
Cell Permeable
Specifications
Specifications
Potency: 840μg/mg
Hygroscopic
DNA synthesis, RNA synthesis
C27H29NO10·HCl
H2O
Red crystalline solid
5mg
23541-50-6
564g/mol