Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Demethylasterriquinone B1
A cell-permeable unsymmetrical bis-indolylquinone insulin mimic with anti-diabetic activity in animal models.
Supplier: MilliporeSigma™ 2600105MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Protect from light | |
Solid | |
95% by HPLC | |
78860-34-1 | |
506.6g/mol |
5 mg | |
Bluish-black | |
Insulin receptor (IR) tyrosine kinase activity | |
C32H30N2O4 | |
DMSO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title