Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Deoxygalactonojirimycin, Hydrochloride
Potent and selective α-galactosidase inhibitor.
Supplier: MilliporeSigma™ 2595445MG
Description
Potent and selective α-galactosidase inhibitor.Specifications
Hygroscopic | |
5 mg | |
98% by TLC | |
75172-81-5 | |
199.6g/mol |
Crystalline Solid | |
White | |
α-glactosidase | |
C6H13NO4·HCl | |
H2O |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title