Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DL-α-Difluoromethylornithine, Hydrochloride
A cell-permeable, anticancer agent that acts as a specific and irreversible inhibitor of ornithine decarboxylase (ODC), the rate-limiting enzyme in polyamine biosynthesis.
Supplier: MilliporeSigma™ 28850025MG
Description
Cell PermeableSpecifications
Hygroscopic | |
25 mg | |
99% by TLC | |
96020-91-6 | |
236.7g/mol |
Powder | |
White | |
ODC | |
C6H12F2N2O2·HCl·H2O | |
MeOH or H2O |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title