Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DNA-PK Inhibitor II
Click to view available options
Quantity:
5 mg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Description | 95% by HPLC |
Target Proteins | DNA-PK |
CAS | 154447-35-5 |
Molecular Formula | C17H15NO3 |
Formula Weight | 281.3g/mol |
Solubility | DMSO or MeOH |
Content And Storage | Protect from light |
Form | Crystalline Solid |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction