Learn More
MilliporeSigma™ Calbiochem™ DNA-PK Inhibitor III
The DNA-PK Inhibitor III, also referenced under CAS 404009-40-1, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 2609621MG
Catalog No. 26-096-21MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Specifications
Specifications
95% by HPLC
Protect from Light
DNA-PK
C12H15NO3
DMSO
Off-white solid
1mg
404009-40-1
221.3g/mol