Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ DNA-PK Inhibitor V
The DNA-PK Inhibitor V, also referenced under CAS 404009-46-7, controls the biological activity of DNA-PK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Supplier: MilliporeSigma™ 2609641MG
Description
Packaged Under Inert GasSpecifications
Yellow | |
DNA-PK | |
C17H17NO3 | |
DMSO | |
Solid |
95% by HPLC | |
404009-46-7 | |
283.3g/mol | |
Protect from light | |
1 mg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction