Learn More
MilliporeSigma™ Calbiochem™ Drosophila Antennapedia Homeo-Domain (43-58)
The membrane translocation signal sequence from Drosophila Antennapedia homeo-domain (43-58) that is reported to act as an inhibitor of protein kinase G Iα (Ki = 970 nM).
Manufacturer:  MilliporeSigma™ 2878951MG
Catalog No. 28-789-51MG
Add to cart
Description
Description
  • Packaged Under Inert Gas
  • Peptide Sequence: H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-OH
Specifications
Specifications
Supplied as a trifluoroacetate salt.
Lyophilized
1mg
C104H168N34O20S
DMSO
95% by HPLC
Hygroscopic
PKG1α
2246.8g/mol