Learn More
EMD Millipore Calbiochem™ Eg5 Inhibitor III, Dimethylenastron Available on GSA/VA Contract for Federal Government customers only.
The Eg5 Inhibitor III, Dimethylenastron, also referenced under CAS 863774-58-7, controls the biological activity of Eg5. This small molecule/inhibitor is primarily used for Membrane applications.
Manufacturer:  EMD Millipore 3246221MG
 View more versions of this product
Catalog No. 32-462-21MG
  Check Availability
Add to cart
Specifications
Description
  • Packaged Under Inert Gas
  • Cell Permeable
Description & Specifications
Specifications
95% by HPLC
Protect from Light
Microtubule-stimulated ATPase activity of the mitotic motor
C16H18N2O2S
DMSO or MeOH
Light yellow solid
1mg
863774-58-7
302.4g/mol