Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ eIF2-GTP-tRNAiMet ternary complex Inhibitor, BTdCPU
Click to view available options
Quantity:
10 mg
Description
A cell-permeable N, N'-diarylurea that acts as an inhibitor of the formation of the ternary complex of eukaryotic translation initiation factor 2 (eIF2), GTP and the initiator methionine tRNA (Met-tRNAi), a key regulatory step of translation initiation. It directly activates heme-regulated inhibitor kinase (HRI), which then phosphorylates eIF2α in cells at 10μM and in cell-free extracts dose-dependently, and thereby prevents respective ternary complex formation without elevating oxidative stress. The sensitivities of various cancer cell lines to the anti-proliferative effects of this compound are shown to correlate with the expression of HRI. Furthermore, this compound displays tumor growth inhibition in a mice xenograft model of MCF-7 human breast cancer cells at 175mg/kg, in vivo .

Specifications
Specifications
Color | White |
Description | 99% by HPLC |
CAS | 1257423-87-2 |
Molecular Formula | C13H8Cl2N4OS |
Formula Weight | 339.2g/mol |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Solid |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction