Learn More
MilliporeSigma™ Calbiochem™ Fostriecin, Sodium Salt, Streptomyces pulveraceous
An antitumor antibiotic that acts as a potent protein phosphatase 2A (PP2A) inhibitor (IC50 = 3.2 nM).
Manufacturer:  MilliporeSigma™ 34428010UG
Catalog No. 34-428-010UG
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
Solid
Colorless
PP2a
C19H26O9PNa
EtOH, MeOH, or H2O
10 μg
98% by HPLC
87860-39-7
452.4g/mol