Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Fumonisin B1, Fusarium moniliforme
Click to view available options
Quantity:
1 mg
Description
Cell Permeable

Specifications
Specifications
Color | White to Beige |
Description | 95% by TLC |
Target Proteins | ceramide synthase |
CAS | 116355-83-0 |
Molecular Formula | C34H59NO15 |
Formula Weight | 721.8g/mol |
Solubility | MeOH |
Form | Powder |
Quantity | 1 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction