Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Gap Junction Enhancer, PQ7
Click to view available options
:
10mg
Description
A cell-permeable quinoline derived compound with low toxicity that enhances gap junctional activity by about 16-fold (at ∽500nM) in T47D breast cancer cells and thereby potentiates the effect of chemotherapeutic agents. Blocks T47D cell colony formation (IC50 = 100nM) and reduce the growth of T47D xenograft tumors by 100% in Nu/Nu mice. Increases caspase-3, - 8, and -9 expression by 1.6, 2.8, and 3.8-fold, respectively to induce apoptotic cell death. Can work synergistically with cisplatin to reduce or totally eliminate tumor growth. Increases the expression of connexins 26, 32, and 43 to allow more efficient trafficking of cisplatin.

Specifications
Specifications
Color | Tan |
Description | 98% by HPLC |
Molecular Formula | C23H19F3N2O3 |
Solubility | DMSO |
Content And Storage | Protect from light |
Form | Powder |
Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction