Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ GIRK Activator, ML297
A blood-brain barrier-permeable, non-toxic phenyl-pyrazolylurea compound that acts as a direct, potent, fast, and reversible activator of GIRK1 (G-protein activated inward-rectifying K+ channel containing subunit 1) containing channels (EC50 = 162, 914, and 887nM in Thallium influx assay for GIRK1/2, GIRK1/3, and GIRK1/4 expressed in HEK-293 cell
Supplier: MilliporeSigma™ 5.05820.0001
Description
A blood-brain barrier-permeable, non-toxic phenyl-pyrazolylurea compound that acts as a direct, potent, fast, and reversible activator of GIRK1 (G-protein activated inward-rectifying K+ channel containing subunit 1) containing channels (EC50 = 162, 914, and 887nM in Thallium influx assay for GIRK1/2, GIRK1/3, and GIRK1/4 expressed in HEK-293 cell lines). Its action does not require the presence of an activated Gi GPCR. Shown to be inactive towards GIRK2, GIRK2/3, Kir2.1, KV7.4 and GABAA, and weakly active against a panel of 61 other receptors, ion channels, enzymes, transporters, and proteins even at higher concentration (∽10μM). Exhibits desirable pharmacokinetic properties with good solubility (17.5μM), predicted hepatic clearance (88 ml/min/kg), and Tmax of 640nM and 130nM in plasma and brain, respectively. Shown to reduce locomotor function and seizure frequency in electroshock- and chemically-induced murine epilepsy models (60mg/kg, i.p).Specifications
Powder | |
10 mg | |
98% by HPLC | |
C17H14F2N4O |
Protect from light | |
White | |
GIRK1/Kir3.1-containing tetrameric GIRK channels | |
DMSO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title