Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ GYKI 52466 Hydrochloride

Manufacturer:  MilliporeSigma™ 5.04973.0001

Catalog No. 50-497-30001


Add to cart

Description

Description

Deactivate with 5% Bleach
Specifications

Specifications

98% by HPLC
Protect from Light
AMPA
329.78g/mol
Yellow solid
10mg
C17H15N3O2·HCl
H2O
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit