Learn More
MilliporeSigma™ Calbiochem™ H-Gly-Arg-Ala-Asp-Ser-Pro-OH
Inactive control peptide for other fibronectin inhibitors
Supplier:  MilliporeSigma™ 033400525MG
 View more versions of this product
Catalog No. 03-340-0525MG
Add to Cart
Description
Description
Specifications
Specifications
≥98% by HPLC
-20°C
Supplied as a trifluoroacetate salt
5 mg
White to Off-white
Lyophilized Solid
H-Gly-Arg-Ala-Asp-Ser-Pro-OH
H-Gly-Arg-Ala-Asp-Ser-Pro-OH
Product Suggestions
Product Suggestions