Learn More
MilliporeSigma™ Calbiochem™ HDAC Inhibitor XVII
A cell-permeable N-hydroxycinnamide derivative that acts a highly potent and HDAC8-selective histone deacetylase inhibitor (IC50 = 27nM), displaying much reduced or little potency against HDAC1/3 (IC50
Supplier: MilliporeSigma™ 5.00671.0001
Description
A cell-permeable N-hydroxycinnamide derivative that acts a highly potent and HDAC8-selective histone deacetylase inhibitor (IC50 = 27nM), displaying much reduced or little potency against HDAC1/3 (IC50 = 3.0μM), HDAC2/4/6/10/11 (IC50 >20μM), or total HDAC activity in HeLa nuclear extract (IC50 >10μM). Reported to preferentially inhibit the proliferation of human lung cancer cells CL1-5, H1299, and A549 (IC50 from 7 to 8μM), while being much less toxic to normal human lung IMR-90 cells (73% inhibition at 40μM).Specifications
Protect from light | |
Powder | |
99% by HPLC | |
C22H19NO3 | |
DMSO |
10 mg | |
Light Beige | |
HDAC8 selective | |
345.39g/mol |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.