EMD Millipore™ Calbiochem™ Horseradish Peroxidase

Suitable for conjugation to proteins and antibodies and as cytochemical marker in immunohistochemical staining

Manufacturer: EMD Millipore™ 51653125KU

 View more versions of this product

Catalog No. 51-653-125KU

  N/A
Add to Cart

Disclaimers:

For Research Use Only

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Quantity 25KU
    Format Lyophilized solid
    Product Type Horseradish Peroxidase
    Storage Requirements -20°C

    Note: 1KU = 1000 units.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit