Learn More
MilliporeSigma™ Calbiochem™ Human Heregulin, Recombinant, E. coli
For cell culture applications
Manufacturer:  MilliporeSigma™ PF04850UG
Catalog No. PF04850UG
Add to cart
Description
Description
  • Shown to be mitogenic for Schwann cells and various epithelial cells
  • Recommended that protein be titrated for optimal results in individual systems
Specifications
Specifications
Lyophilized
≥97% by SDS-PAGE
-20°C
Human Heregulin, Recombinant, E. coli
Lyophilized from PBS, 50μg BSA/μg cytokine.
50μg
Neu Differentiation Marker, NDF