Learn More
Specifications
Specifications
| Formulation | Lyophilized from 150mM NaCl, 20mM phosphate buffer, pH 7.4 |
| Quantity | 10 mg |
| Test Specificity | Nonreactive for HBsAg and for HCV, HBc, and HIV antibodies |
| Content And Storage | -20°C |
| Form | Lyophilized |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.