Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Human Plasma Fibronectin
Promotes attachment of cells to commonly used culture substrates.
$269.00 - $956.00
Specifications
Content And Storage | -20°C |
---|---|
Product Type | Human Plasma Fibronectin |
Formulation | In PBS, pH 7.3. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
34-163-55MG
|
MilliporeSigma™
3416355MG |
5 mg |
Each for $956.00
|
|
|||||
34-163-51MG
|
MilliporeSigma™
3416351MG |
1 mg |
Each for $269.00
|
|
|||||
Description
EMD Millipore Calbiochem™ Human Plasma Fibronectin is purified from human plasma.
- Native fibronectin
- Contaminants: None
- Detected by IEP with antiserum to fibronectin or normal human serum
- Single Major B by SDS-PAGE
Specifications
-20°C | |
Human Plasma Fibronectin | |
In PBS, pH 7.3. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title