Learn More
MilliporeSigma™ Calbiochem™ Human Recombinant BMP-1/PCP, CF
Cleaves fluorogenic peptide substrate
Supplier:  MilliporeSigma™ 20363810UG
Catalog No. 20-363-810UG
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Consists of amino acids 121to730
  • Expressed in NS0-derived mouse myeloma cells with C-terminal His-Tag™ sequence
Specifications
Specifications
Solid
≥95% by SDS-PAGE
-20°C
>2 pmol/min/μg as determined by cleavage of a fluorogenic peptide substrate, Mca-YVADAPK (Dnp)-OH
Lyophilized from 0.2μm filtered HEPES, NaCl, CaCl2, ZnCl2, and Brij™ 35 detergent.
10μg
Procollagen C-proteinase, PCP, Human bone morphogenetic protein (BMP-1)
Human Recombinant BMP-1/PCP, CF