EMD Millipore™ Calbiochem™ Hygromycin B, Streptomyces sp. Sterile-Filtered Soln in 25mM HEPES Available on GSA/VA Contract for Federal Government customers only.

For identification or selection of recombinant clones in variety of cell types

Manufacturer: EMD Millipore™ 4000535ML

 View more versions of this product

Catalog No. 40-005-35ML

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Quantity 5mL
    Storage Requirements 2- 8°C
    Product Type Hygromycin B Sterile-Filtered Solution in 25mM HEPES
    Concentration 50μg/mL to 1mg/mL for mammalian cell selection
    Format Liquid

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit