Learn More
EMD Millipore™ Calbiochem™ Hygromycin B, Streptomyces sp. Sterile-Filtered Solution Available on GSA/VA Contract for Federal Government customers only.
For identification or selection of recombinant clones in variety of cell types
Manufacturer:  EMD Millipore™ 4000525ML
 View more versions of this product
Catalog No. 40-005-25ML
  Check Availability
Add to cart
Disclaimers:
For Research Use Only
Specifications Description & Specifications
Specifications
50μg/mL to 1mg/mL for mammalian cell selection
Hygromycin B Sterile-Filtered Solution
2- 8°C
Liquid
5mL