Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Hygromycin B, Streptomyces sp. Sterile-Filtered Solution Available on GSA/VA Contract for Federal Government customers only.

For identification or selection of recombinant clones in variety of cell types

$70.00 - $389.00

Specifications

Product Type Hygromycin B Sterile-Filtered Solution
Storage Requirements 2- 8°C
Format Liquid
Concentration 0.05 to 1mg/mL for mammalian cell selection
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
40-005-25ML
View Documents
MilliporeSigma™
4000525ML
5mL Each for $70.00
Add to cart
 
40-005-220ML
View Documents
MilliporeSigma™
40005220ML
20mL Each for $205.00
Add to cart
 
40-005-250ML Available on GSA/VA Contract for Federal Government customers only.
View Documents
MilliporeSigma™
40005250ML
50mL Each for $389.00
Add to cart
 
Description

Description

  • Aminoglycoside antibiotic
  • Inhibits growth of prokaryotic (bacteria) and eukaryotic microorganisms (yeasts) and mammalian cells
  • Inhibits protein synthesis at translocation step on 70s ribosome
  • Penetrates cells that have been permeabilized by virus infection
  • Causes misreading of mRNA
  • Can act as an effective antiviral agent

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

Hygromycin B Sterile-Filtered Solution
2- 8°C
Liquid
0.05 to 1mg/mL for mammalian cell selection

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit