Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ IL-1R Antagonist
The IL-1R Antagonist, also referenced under CAS 566914-00-9, controls the biological activity of IL-1R.
Supplier: MilliporeSigma™ 40761610MG
Description
- Packaged Under Inert Gas
- Cell Permeable
Specifications
Colorless | |
TIR domain-mediated MyD88/IL1-RI interaction | |
C18H26N2O2 | |
DMSO or EtOH | |
10 mg |
95% by HPLC | |
566914-00-9 | |
302.4g/mol | |
Oil |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction