Learn More
MilliporeSigma Calbiochem InhibitorSelect mTOR Signaling Regulators Panel
A panel containing 17 potent and selective small molecule regulators that is useful for the study of mTOR signaling: 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat
Manufacturer: MilliporeSigma 4759951EA
Description
A panel containing 17 potent and selective small molecule regulators that is useful for the study of mTOR signaling: 1 mg of Akt Inhibitor VIII, Isozyme-Selective, Akti-1/2 (Cat. No. 124018), 10 mg of Akt Inhibitor XVI, AT7867 (Cat. No. 124033), 1 mg of AMPK Inhibitor, Compound C (Cat. No. 171260), 10 mg of bpV(phen) (Cat. No. 203695), 5 mg of Compound 401 (Cat. No. 234501), 1 mg of H-89, Dihydrochloride (Cat. No. 371963), 5 mg of mTOR Inhibitor III, PP242 (Cat. No. 475988), 5 mg of mTOR Inhibitor IV, Ku-63794 (Cat. No. 475990), 5 mg of PDK1 Inhibitor II, BX-517 (Cat. No. 521276), 1 mg of PI-103 (Cat. No. 528100), 1 mg of PI 3-Kβ Inhibitor VI, TGX-221 (Cat. No. 528113), 5 mg of PTK/PI 3-K/mTOR Inhibitor, PP121 (Cat. No. 529584), 1 mg of Rapamycin (Cat. No. 553210), 10 mg of S6K1 Inhibitor, PF-4708671 (Cat. No. 559273), 10 mg of SHIP1 Inhibitor, 3AC (Cat. No. 565835), 10 mg of SHIP2 Inhibitor, AS1938909 (Cat. No. 565840), 1 mg of U0126 (Cat. No. 662005). Also provided with 15mL of anhydrous DMSO (Cat. No. KP31817).Specifications
Solid | |
Protect from Light | |
1 ea | |
DMSO |