Learn More
MilliporeSigma™ Calbiochem™ InhibitorSelect™ TLR Signaling Pathway Panel
A panel containing 16 potent, selective and cell-permeable inhibitors that is useful for the study of TLR signaling: 1 mg of TIRAP Inhibitor Peptide, Cell-Permeable (Cat. No. 613570), 1
Supplier: MilliporeSigma™ 5.00528.0001
Description
A panel containing 16 potent, selective and cell-permeable inhibitors that is useful for the study of TLR signaling: 1 mg of TIRAP Inhibitor Peptide, Cell-Permeable (Cat. No. 613570), 10 mg of IL-1R Antagonist (Cat. No. 407616), 5 mg of Interleukin-1 Receptor-Associated-Kinase-1/4 Inhibitor (Cat. No. 407601), 1 mg of (5Z)-7-Oxozeaenol, Curvularia sp. (Cat. No. 499610), 10 mg of BX-795 (Cat. No. 204001), 5 mg of NBD-Binding Peptide II, Cell Permeable, CTP-NBD (Cat. No. 480026), 25 mg of Cdc42/Rac1 GTPase Inhibitor, ML141 (Cat. No. 217708), 5 mg of PKR Inhibitor (Cat. No. 527450), 5 mg of LY294002 (Cat. No. 440202), 5 mg of U0126 (Cat. No. 662005), 1 mg of IKK Inhibitor VII (Cat. No. 401486), 10 mg of BAY 11-7082 (Cat. No. 196870), 1 mg of SB 203580 (Cat. No. 559389), 5 mg of SP600125 (Cat. No. 420119), 10 mg of STAT3 Inhibitor III, WP1066 (Cat. No. 573097), and 10 mg of MG-132 (Cat. No. 474787). Also provided with 15mL of anhydrous DMSO (Cat. No. KP31817).Specifications
Major hubs involved in TLR signaling cascade | |
Hygroscopic, Protect from light | |
Solid | |
1 Each |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.