Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma Calbiochem Novobiocin, Sodium Salt
Novobiocin, Sodium Salt, CAS 1476-53-5, is a DNA gyrase inhibitor that is used for the production of positively supercoiled plasma DNA. Inhibits retrovirus RNA-dependent DNA polymerases.
Manufacturer: MilliporeSigma 49120710GM
Description
DNA gyrase inhibitor used for the production of positively supercoiled plasma DNA. Inhibits retrovirus RNA-dependent DNA polymerases. Also a potent inhibitor of ADP ribosylation eliminates the wild-type pMG110 plasmid from E. coli .Specifications
90% by HPLC | |
Protect from Light | |
retrovirus RNA-dependent DNA polymerases | |
C31H35N2O11·Na | |
H2O |
White to off-white solid | |
10gm | |
1476-53-5 | |
634.6g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title