Learn More
MilliporeSigma™ Calbiochem™ Novobiocin, Sodium Salt
Novobiocin, Sodium Salt, CAS 1476-53-5, is a DNA gyrase inhibitor that is used for the production of positively supercoiled plasma DNA. Inhibits retrovirus RNA-dependent DNA polymerases.
Manufacturer:  MilliporeSigma™ 4912071GM
 View more versions of this product
Catalog No. 49-120-71GM
Add to cart
Description
Description
DNA gyrase inhibitor used for the production of positively supercoiled plasma DNA. Inhibits retrovirus RNA-dependent DNA polymerases. Also a potent inhibitor of ADP ribosylation eliminates the wild-type pMG110 plasmid from E. coli .
Specifications
Specifications
90% by HPLC
Protect from Light
retrovirus RNA-dependent DNA polymerases
C31H35N2O11·Na
H2O
White to off-white solid
1gm
1476-53-5
634.6g/mol