Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ Bafilomycin A1, ≥97% by HPLC
Click to view available options
Quantity:
15 μg
Description
- Packaged Under Inert Gas
- Cell Permeable

Specifications
Specifications
Formulation | A 100μM (15μg/241μL) sterile-filtered solution of Bafilomycin A1, 97% by HPLC in 90% DMSO. |
Description | 97% by HPLC |
Target Proteins | vacuolar-type H+-ATPase |
CAS | 88899-55-2 |
Molecular Formula | C35H58O9 |
Formula Weight | 622.8g/mol |
Content And Storage | Avoid Freeze-Thaw Cycles, Hygroscopic, Protect from Light |
Form | Liquid |
Quantity | 15 μg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction