Learn More
MilliporeSigma™ Calbiochem™ InSolution™ cFMS Receptor Tyrosine Kinase Inhibitor
The InSolution™ cFMS Receptor Tyrosine Kinase Inhibitor controls the biological activity of cFMS Receptor Tyrosine Kinase. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Manufacturer:  MilliporeSigma™ 3440411MG
Catalog No. 34-404-11MG
Add to cart
Description
Description
Packaged Under Inert Gas
Specifications
Specifications
Liquid
A 10 mM (1mg/273μl) solution of cFMS Receptor Tyrosine Kinase Inhibitor in DMSO.
C20H22N4O3
1 mg
95% by HPLC
366.4g/mol