Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ MPS1 Inhibitor, NMS-P715
A 5 mM (2 mg/592 μl) sterile-filtered solution of MPS1 Inhibitor, NMS-P715 in DMSO.
Manufacturer: MilliporeSigma™ 5.06313.0001
Description
- Packaged Under Inert Gas
- Cell Permeable
- Deactivate with 5% Bleach
Specifications
A 5 mM (2mg/592μl) sterile-filtered solution of MPS1 Inhibitor, NMS-P715 in DMSO. | |
Liquid | |
2mg | |
C35H39F3N8O3 |
98% by HPLC | |
Avoid Freeze-Thaw Cycles Hygroscopic Protect from Light |
|
MPS1 | |
676.3g/mol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title