Learn More
MilliporeSigma Calbiochem InSolution Okadaic Acid, Prorocentrum sp.
InSolution Okadaic Acid, Prorocentrum sp. CAS 78111-17-8, is a 250µM solution of Okadaic Acid in DMSO. Potent inhibitor of protein phosphatase 1 (IC50 = 10-15nM) and 2A (IC50 = 0.1nM).
Manufacturer: MilliporeSigma 49560925UG
Description
InSolution Okadaic Acid, Prorocentrum sp. CAS 78111-17-8, is a 250µM solution of Okadaic Acid in DMSO. Potent inhibitor of protein phosphatase 1 (IC50 = 10-15nM) and 2A (IC50 = 0.1nM).Specifications
A 250μM (25μg/124μl) solution of Okadaic Acid, in DMSO. | |
Liquid | |
25μg | |
C44H68O13 |
95% by HPLC | |
Hygroscopic Protect from Light |
|
protein phosphatase | |
805g/mol |