Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ InSolution™ Rapamycin
InSolution™ Rapamycin, CAS 53123-88-9, is a 5 mM (500 μg/109 μl) solution of Rapamycin (Cat. No. 553210) in DMSO. Selectively inhibits mTOR and blocks activation of p70 S6 Kinase (IC50 = 50 pM).
Supplier: MilliporeSigma™ 553211500UG
Description
Packaged Under Inert GasSpecifications
A 5 mM (500μg/109μl) solution of Rapamycin in DMSO. | |
Mammalian target of rapamycin (mTOR) | |
914.2g/mol | |
Liquid |
98% by TLC | |
C51H79NO13 | |
Hygroscopic, Protect from light | |
500 μg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction