Learn More
EMD Millipore Calbiochem™ InSolution™ Rapamycin Available on GSA/VA Contract for Federal Government customers only.
InSolution™ Rapamycin, CAS 53123-88-9, is a 5 mM (500 µg/109 µl) solution of Rapamycin (Cat. No. 553210) in DMSO. Selectively inhibits mTOR and blocks activation of p70 S6 Kinase (IC50 = 50 pM).
Manufacturer:  EMD Millipore 5532111MG
 View more versions of this product
Catalog No. 55-321-11MG
  Check Availability
Add to cart
Specifications
Description
Packaged Under Inert Gas
Description & Specifications
Specifications
A 5 mM (500μg/109μl) solution of Rapamycin in DMSO.
Liquid
1mg
C51H79NO13
98% by TLC
Hygroscopic
Protect from Light
Mammalian target of rapamycin (mTOR)
914.2g/mol