Learn More
MilliporeSigma™ Calbiochem™ iPSC Induction Enhancer II, OAC1
A cell-permeable pyrrolopyridinyl-benzamide compound that is reported to enhance the efficiency of 4F/4TF-induced iPSC generation from MEFs cultured in either feeder cell-containing ESC media or in feeder-/serum-free iSF1 media (1μM OAC1 treatment starts 48 h after viral transduction)
Supplier:  MilliporeSigma™ 5.00501.0001
Catalog No. 50-050-10001
Add to Cart
Description
Description
A cell-permeable pyrrolopyridinyl-benzamide compound that is reported to enhance the efficiency of 4F/4TF-induced iPSC generation from MEFs cultured in either feeder cell-containing ESC media or in feeder-/serum-free iSF1 media (1μM OAC1 treatment starts 48 h after viral transduction). OCA1 is shown to increase cellular Oct4, Nanog, Sox2, and Tet1 mRNA levels via a yet unidentified mechanism, although OAC1 is reported not affect Oct4 promoter methylation, cellular Wnt signaling, or p53 and p21 mRNA levels. Six other iPSC reprogramming improving reagents, AM580, AzaC, BIX-01294 (Cat. No. 382190), Tranylcypromine (Cat. No. 616431), VPA (Cat. No. 676380), and Vitamin C (Cat. No. 1813), in comparison, do not exhibit Oct4 and Nanog promoters enhancing activity, while BIO (Cat. No. 361550) is demonstrated to be 3.8- and 6-fold less potent than OAC1, respectively, in inducing Oct4 and Nanog transcription.
Specifications
Specifications
Protect from light
Solid
99% by HPLC
237.3g/mol
25 mg
Off-white
C14H11N3O
DMSO